Supelco® Organic Reference Standard, EPA 8270 Organophosphorus Pesticide Mix 2
CRM47908
: Merck
:
- Concentration:2000 µg/ml each component in dichloromethane
- Pk:1
- Standards:Certified reference material
- Description:EPA 8270 Organophosphorus pesticide mix 2
- Storage conditions:2…8 °C
- Hazard class description:H301 + H311 - H315 - H319 - H330 - H336 - H351 - H410
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Pk Info:Ampule of 1 ml
- GHS signal: GHS06, GHS08, GHS09
: P202 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310 - P305 + P351 + P338