Order Entry
Netherlands
ContactUsLinkComponent
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥98% (by TLC) fluorescent pH indicator
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥98% (by TLC) fluorescent pH indicator
Catalog # 90041.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥98% (by TLC) fluorescent pH indicator
Catalog # 90041.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :557.467 g/mol
Storage Temperature:Freezer
CAS Number:150347-59-4
 :MFCD31812843
Formula:C₂₉H₁₉NO₁₁
MDL Number:MFCD00037466