Supelco® Organic Reference Standard, Furosemide Related Compound A
PHR1817-100MG
: Merck
:
- Pk:100 mg
- Standards:Pharmaceutical Secondary Standard; Certified Reference Material
- CAS No.:4818-59-1
- Description:Furosemide related compound A
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD18379506
- M-Clarity quality level:MQ300
- Synonym(s):2-Chloro-4-furfurylamino-5-sulfamoylbenzoic acid; 2-CHLORO-4-N-FURFURYLAMINO -5-SULFAMOYLBENZOIC ACID
- Melting point:197 °C (dec.) (lit.)
- Molecular weight:330,74