Order Entry
Netherlands
ContactUsLinkComponent
5-Carboxy-X-rhodamine-PEO₈-propionate succinimidyl ester amine-reactive fluorescent dye
5-Carboxy-X-rhodamine-PEO₈-propionate succinimidyl ester amine-reactive fluorescent dye
Catalog # 90089.
Supplier:  Biotium
CAS Number:  
undefined
5-Carboxy-X-rhodamine-PEO₈-propionate succinimidyl ester amine-reactive fluorescent dye
Catalog # 90089.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1055.17 g/mol
Storage Temperature:Freezer
Formula:C₅₆H₇₀N₄O₁₆