6-(Fluorescein-6-carboxamido) hexanoic acid succinimidyl ester (6-SFX) amine-reactive fluorescent dye
Catalog # 90096.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:586.55 g/mol
Storage Temperature:Freezer
:148356-00-7
Formula:C₃₁H₂₆N₂O₁₀