5(6)-Carboxy-X-rhodamine triethylammonium salt ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90004.
Supplier: Biotium
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
- Environmentally Preferable:
Specifications
About this item
Synonyms: Carboxy-X-rhodamine triethylammonium salt, 5 and 6 isomers, 5(6)-ROX)
:
:635.6 g/mol
Storage Temperature:Freezer
:198978-94-8
Formula:C₃₉H₄₅N₃O₅