Supelco® Organic Reference Standard, Salbutamol Impurity B
PHR1959-50MG
: Merck
:
- Pk:50 mg
- Standards:Pharmaceutical Secondary Standard; Certified Reference Material
- CAS No.:96948-64-0
- Description:Salbutamol impurity B
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):(1RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxyphenyl)ethanol; Albuterol Related Compound I; 2-tert-Butylamino-1-(4-hydroxyphenyl)ethanol
- Molecular weight:209,28
- GHS signal: GHS07
: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338