Supelco® Organic Reference Standard, Allopurinol Related Compound F
Catalog # PHR1981-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Pharmaceutical Secondary Standard; Certified Reference Material
- CAS No.:321571-07-7
- Description:Allopurinol related compound F
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):Ethyl (E/Z)-3-(2-carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate
- Molecular weight:278,26
Specifications
About this item
- GHS signal: GHS07
Caution: P305 + P351 + P338