Supelco® TraceCERT® Organic Reference Standard, Ethyl methanesulfonate
72945-120MG
: Merck
:
- Pk:120 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:62-50-0
- Description:Ethyl methanesulfonate
- Storage conditions:2…8 °C
- Hazard class description:H302 - H315 - H317 - H319 - H340 - H350 - H361f
- Shipping conditions:STD
- MDL No.:MFCD00007559
- M-Clarity quality level:MQ300
- Synonym(s):Ethyl mesylate; Methanesulfonic acid ethyl ester
- Formula:CH₃SO₃CH₂CH₃
- Molecular weight:124,16
- Density:1,206 g/ml at 20 °C
- Flash point:100 °C
- Boiling point:85 - 86 °C/10 mmHg (lit.)
- GHS signal: GHS08, GHS07
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P202 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338 - P308 + P313