Supelco® Organic Reference Standard, Ethyl Benzenesulfonate
13295-120MG
: Merck
:
- Pk:120 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:515-46-8
- Description:Ethyl benzenesulfonate
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00025033
- M-Clarity quality level:MQ300
- Synonym(s):Benzenesulfonic acid ethyl ester; Ethyl phenylsulfonate; NSC 3217
- Molecular weight:186,23
- Flash point:Not applicable
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P264 - P270 - P301 + P312 - P501