Supelco® Drug and metabolite standards, Valproic acid solution, (CRM)
Catalog # V-006-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:99-66-1
- Description:Valproic acid solution
- Description:Valproic acid solution
- Pk Info:ampoule of 1 ml