Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, methedrone hydrochloride solution, (CRM)
  M-147-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, methedrone hydrochloride solution, (CRM)
  M-147-1ML
 :  Merck
 :  

 

  • Concentration:
    1,0 mg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    879665-92-6
  • Description:
    Methedrone hydrochloride solution
  • Description:
    Methedrone hydrochloride solution
  • Pk Info:
    ampoule of 1 ml