Supelco® Drug and metabolite standards, Cyclobenzaprine-D3 hydrochloride solution, (CRM)
Catalog # C-114-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:118983-42-3
- Description:Cyclobenzaprine-D3 hydrochloride solution
- Environmentally Preferable:
- Description:Cyclobenzaprine-D3 hydrochloride solution
- Pk Info:ampoule of 1 ml