Supelco® Drug and metabolite standards, Aripiprazole-d8 solution, (CRM)
Catalog # A-081-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1089115-04-7
- Description:Aripiprazole-d8 solution
- Description:Aripiprazole-d8 solution
- Pk Info:ampoule of 1 ml