Supelco® Cerilliant® Organic Reference Standard, 25-Hydroxyvitamin D2 solution
Catalog # H-073-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:50 µg/ml in ethanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:21343-40-8
- Description:25-Hydroxyvitamin D2 solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H319
- Shipping conditions:DICE
- MDL No.:MFCD02683431
- M-Clarity quality level:MQ300
- Synonym(s):25-Hydroxyvitamin D2 solution
- Molecular weight:412,65
- Flash point:14 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338