Order Entry
Netherlands
ContactUsLinkComponent
Safety spectacles, Polarvision
  UVEX9231.960
 :  UVEX
undefined
Safety spectacles, Polarvision
  UVEX9231.960
 :  UVEX
  • Type:
    Anti-UV, anti-scratch
  • Colour:
    Black/white
  • Environmentally Preferable:
  • Lenses:
    Grey PC
  • Pk:
    1
Comfortable safety spectacles with polarisation filter.
  • Polarisation filter protects against reflections and glare (e.g. water surfaces and highly polished materials)
  • Scratch-resistant coated
Suitable for workstations with reflective surfaces (offshore, maritime, drivers, construction industry).