Order Entry
Netherlands
ContactUsLinkComponent
Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Paraplast® Plus
Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Paraplast® Plus
Catalog # LEIC8889502004
Supplier:  LEICA MICROSYSTEMS
CAS Number:  
Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Paraplast® Plus
Catalog # LEIC8889502004
Supplier:  LEICA MICROSYSTEMS
CAS Number:  

Specifications

  • Pk:
    8 kg
  • Seq:
    4138

Specifications

About this item

 :
 :1
CAS Number:8002-74-2
 :8012-95-1