Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, 2-Butanone-2,4-dinitrophenylhydrazone, Dr Ehrenstorfer
Organic reference standard, 2-Butanone-2,4-dinitrophenylhydrazone, Dr Ehrenstorfer
Catalog # EHERC10862050
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2-Butanone-2,4-dinitrophenylhydrazone, Dr Ehrenstorfer
Catalog # EHERC10862050
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Standards:
    Reference standard
  • CAS No.:
    958-60-1
  • Description:
    2-Butanone-2,4-dinitrophenylhydrazone
  • Formula:
    C₁₀H₁₂N₄O₄
  • Description:
    2-Butanone-2,4-dinitrophenylhydrazone

Specifications

About this item

Certifications: Produced in accordance to ISO 17034