Flame thermionic detector (FTD)
Flame thermionic detector, FTD-2010 Plus, 230 V
Catalog # SHIM221-73332-48
Supplier: SHIMADZU
Jump to:
Product Details & Documents
This detector features a digital pressure controller and is applicable for GC-2010/2010 Plus.
: Includes one piece of collector as a standard accessory.
: FTD requires hydrogen and air as supply gas. Supply air should be from a high-purity air cylinder. It is not recommended to use air from a compressor.
Specifications
- Description:Flame thermionic detector, FTD-2010 Plus, 230 V
Accessories
Product Information
- Catalog #AvailabilityPrice
- Hex nut screwdriver
Product Information
- Catalog #AvailabilityPrice
- Nozzle (0,3 Ø, standard)
Product Information
- Catalog #AvailabilityPrice
- FTD-2010 Plus/2014C collector