Order Entry
Netherlands
ContactUsLinkComponent
Glass Microfiber Filter Paper
Glass Microfiber Filter Paper
Catalog # PRATA090875
Supplier:  PRAT DUMAS
undefined
Glass Microfiber Filter Paper
Catalog # PRATA090875
Supplier:  PRAT DUMAS

Specifications

  • Grade:
    GF 6
  • Seq:
    03
  • Size:
    90 mm
  • Thickness:
    480 µm
  • Weight:
    75 g/m²
  • Pk:
    100

Specifications

About this item

These glass microfiber filter papers allow fast filtration and high particle retention. They can be used at temperatures up to 380 °C.

  • Chemical compatibility with many solvents and with reagents
  • Suitable for microfiltration