Order Entry
Netherlands
ContactUsLinkComponent
BAC resolution control standard n-P (6 components)
BAC resolution control standard n-P (6 components)
Catalog # REST36010
Supplier:  Restek
CAS Number:  
undefined
BAC resolution control standard n-P (6 components)
Catalog # REST36010
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    100 mg/dl each in water
  • Pk:
    1 mL
  • Description:
    BAC resolution control standard n-P
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mix is use to verify the retention time for each compound normally included in a blood alcohol test, and to verify that the compounds are resolved from and do not interfere with one another.

  • Includes 1-propanol internal standard
  • Intended for qualitative use only

It consists of acetaldehyde, acetone, ethanol (BAC), methanol, 1-propanol (n-propanol) and 2-propanol (isopropanol).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.