508.1 internal standard (pentachloronitrobenzene)
Catalog # REST32091
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 μg/ml in ethyl acetate
- Pk:1
- CAS No.:82-68-8
- Description:508.1 internal standard (pentachloronitrobenzene)
- Formula:C₆Cl₅NO₂
- Description:Pentachloronitrobenzene
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.