VOA calibration mix #2 (7 components)
Catalog # REST30007
Supplier: Restek
CAS Number:
Specifications
- Concentratie:2000 μg/ml each in P and T methanol
- VPE:1
- Omschrijving:VOA calibration mix #2
- Info VPE:1 ml/ampoule
Specifications
About this item
Mix contains: Benzene (71-43-2), Carbon disulfide (75-15-0), Ethylbenzene (100-41-4), Toluene (108-88-3), vinyl acetate (108-05-4), o-Xylene (95-47-6), p-Xylene (106-42-3).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.