ASTM D6042-96 calibration mix (7 components)
Catalog # REST31628
Supplier: Restek
CAS Number:
Specifications
- Concentration:50 μg/ml each in isopropanol
- Pk:1 mL
- Description:ASTM D6042-96 calibration mix
- Pk Info:1 ml/ampoule
Specifications
About this item
This mixture contains the common antioxidants and slips listed in ASTM D6042-96, along with BHT.
The mix consists of BHT, erucamide slip, vitamin E, irgafos 168, irganox 3114, irganox 1010, irganox 1076.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.