Order Entry
Netherlands
ContactUsLinkComponent
 
Tetrapentyltin internal standard
Catalog # REST31475
Supplier:  Restek
CAS Number:  
undefined
Tetrapentyltin internal standard
Catalog # REST31475
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in methylene chloride
  • Pk:
    1 mL
  • CAS No.:
    3765-65-9
  • Description:
    Tetrapentyltin
  • Formula:
    C₂₀H₄₄Sn
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Complete data pack available for audit compliance.