Order Entry
Netherlands
ContactUsLinkComponent
GC multiresidue pesticide standard #1 (16 components)
GC multiresidue pesticide standard #1 (16 components)
Catalog # REST32563
Supplier:  Restek
CAS Number:  
undefined
GC multiresidue pesticide standard #1 (16 components)
Catalog # REST32563
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml each in toluene
  • Pk:
    1 mL
  • Description:
    GC multiresidue pesticide standard #1
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Organophosphorus compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.

Contains: Azinphos ethyl, azinphos methyl, chlorpyrifos, chlorpyrifos methyl, diazinon, EPN, fenitrothion, isazophos, phosalone, phosmet, pirimiphos ethyl, pirimiphos methyl, pyraclofos, pyrazophos, pyridaphenthion, quinalphos.