4,6-Dinitro-2-methylphenol (dinitro-o-cresol)
REST31292
: Restek
:
- Concentration:1000 µg/mL in methanol
- Pk:1 mL
- CAS No.:534-52-1
- Description:4,6-Dinitro-2-methylphenol (dinitro-o-cresol)
- Formula:C₇H₆N₂O₅
- Description:4,6-Dinitro-2-methylphenol (dinitro-o-cresol)
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.