Order Entry
Netherlands
ContactUsLinkComponent
MA volatile petroleum hydrocarbon (VPH) standard (13 components)
Catalog # REST30434
Supplier:  Restek
CAS Number:  
undefined
MA volatile petroleum hydrocarbon (VPH) standard (13 components)
Catalog # REST30434
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    MA volatile petroleum hydrocarbon (VPH) standard, in P and T methanol
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
Standard contains (C5) n-pentane, 1000 µg/ml; (C9) n-nonane, 1000 µg/ml; benzene, 500 µg/mL; ethylbenzene, 500 µg/ml; 2-methylpentane , 1500 µg/ml; methyl tert-butyl ether (MTBE), 1500 µg/ml; naphthalene, 1000 µg/ml; toluene , 1500 µg/ml; 1,2,4-trimethylbenzene , 1000 µg/ml; 2,2,4-trimethylpentane (isooctane) , 1500 µg/ml; m-xylene , 1000 µg/ml; o-xylene, 1000 µg/ml; p-xylene , 1000 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.