Order Entry
Netherlands
ContactUsLinkComponent
Mercury(II) iodide ≥99.0% ACS, Fluka™
Mercury(II) iodide ≥99.0% ACS, Fluka™
Certificates
Mercury(II) iodide ≥99.0% ACS, Fluka™
Certificates

Specification Test Results

  • Assay (excluding dried substances):
    Min. 99.0 %
  • Solubility:
    complying
  • Mercury (Hg⁺):
    Max. 0.1 %
  • Soluble mercury salts (as Hg):
    Max. 0.05 %

Specification Test Results

Related Information

Frequently Bought Together