Bromocresol purple Reag. Ph. Eur. indicator, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:115-40-2
:
Formula:C₂₁H₁₆Br₂O₅S
MDL Number:MFCD00011681
:540.23 g/mol
Merck Index:13,01371
Melting Pt:240 °C (dec.)
Storage Temperature:Ambient
EINECS:204-087-8
Specification Test Results
- Change range:complying
- Indicator solution:complying
- Test of sensitivity:complying
- Curve course (UV):complying
- Extinction maximum at:414 - 424 nm
- Extinction maximum of:Min. 400
- pH Range:5.2 - 6.8, yellow to purple
Specification Test Results
Frequently Bought Together