Order Entry
Netherlands
ContactUsLinkComponent
Barium sulphate 97.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Barium sulphate 97.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Leverancier:  Honeywell Chemicals
Certificaten
Barium sulphate 97.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Leverancier:  Honeywell Chemicals

Specificatie testresultaten

  • Assay:
    97.5 - 100.5 %
  • Loss on ignition (600°C):
    Max. 2.0 %
  • Soluble in acid:
    Max. 0.3 %
  • Soluble Ba-salts:
    complying
  • Acid. or alk. react. impurities:
    complying
  • Oxidizable S compounds:
    complying
  • Residual solvents:
    complying

Specificatie testresultaten

Gerelateerde informatie

Frequently Bought Together