Sodium hydroxide 0.5 M NaOH (0.5N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Density:1.020 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,III
:GHS05
Specification Test Results
- Calibrated Against:potassium hydrogen phthalate
- Appearance of the substance:complying
- Assay:0.499 - 0.501 mol/l
Specification Test Results
Frequently Bought Together