Mouse Recombinant IL-6 (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death.
- High quality
- Low endotoxin
- Affordable
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: B9 cell proliferation, ED50 ≤0,05 ng/ml, bioactivity ≥2,0×10⁷ units/mg
- Gene ID:P08505
- Reconstitution instructions:Sterile 10 mM HCl at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:IFN-β2|HSF|BSF-2|B-cell differentiation factor|MGI-2
- Protein/peptide name:IL6
- Purity:≥95%
- Molecular weight:21,9 kDa
- Sequence:MFPTSQVRRG DFTEDTTPNR PVYTTSQVGG LITHVLWEIV EMRKELCNGN SDCMNNDDAL AENNLKLPEI QRNDGCYQTG YNQEICLLKI SSGLLEYHSY LEYMKNNLKD NKKDKARVLQ RDTETLIHIF NQEVKDLHKI VLPTPISNAL LTDKLESQKE WLRTKTIQFI LKSLEEFLKV TLRSTRQT
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together