Analytical standard, (±)-Metamifop, reference material, Honeywell Fluka™
Catalog # HONCR1326-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:256412-89-2
- Description:(±)-Metamifop
- Formula:C₂₃H₁₈ClFN₂O₄
- Storage temperature:Ambient
- Description:(±)-Metamifop
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results