Order Entry
Netherlands
ContactUsLinkComponent
Analytical standard, Triclabendazole sulfone, reference material, Honeywell Fluka™
Analytical standard, Triclabendazole sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1271-10MG
CAS Number:  
Analytical standard, Triclabendazole sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1271-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    106791-37-1
  • Description:
    Triclabendazole sulfone
  • Melting point:
    190…191 °C
  • Formula:
    C₁₄H₉Cl₃N₂O₃S
  • Storage temperature:
    Ambient
  • Description:
    Triclabendazole sulfone

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results