Order Entry
Netherlands
ContactUsLinkComponent
Analytical standard, Sulfamethizole-(phenyl-¹³C₆), reference material, Honeywell Fluka™
Analytical standard, Sulfamethizole-(phenyl-¹³C₆), reference material, Honeywell Fluka™
  HONCR1245-10MG
 :  
Analytical standard, Sulfamethizole-(phenyl-¹³C₆), reference material, Honeywell Fluka™
  HONCR1245-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1334378-92-5
  • Description:
    Sulfamethizole-(phenyl-¹³C₆)
  • Melting point:
    208 °C
  • Formula:
    ¹³C₆C₃H₁₀N₄O₂S₂
  • Storage temperature:
    Ambient
  • Description:
    Sulfamethizole-(phenyl-¹³C₆)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results