Analytical standard, Clenisohexerol hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1235-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:37158-48-8
- Description:Clenisohexerol hydrochloride
- Melting point:162…166 °C
- Formula:C₁₄H₂₂Cl₂N₂O · HCl
- Storage temperature:Ambient
- Description:Clenisohexerol hydrochloride
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results