Analytical standard, Aldimorph, reference material, Honeywell Fluka™
Catalog # HONCR1200-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:91315-15-0
- Description:Aldimorph
- Melting point:–27…–19 °C
- Formula:C₁₈H₃₇NO (major component)
- Storage temperature:Ambient
- Description:Aldimorph
- Boiling point:110…140 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results