Order Entry
Netherlands
ContactUsLinkComponent
Analytical standard, Didecyldimethylammoni, reference material, Honeywell Fluka™
Analytical standard, Didecyldimethylammoni, reference material, Honeywell Fluka™
Catalog # HONCR1121-250MG
CAS Number:  
Analytical standard, Didecyldimethylammoni, reference material, Honeywell Fluka™
Catalog # HONCR1121-250MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    250 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    7173-51-5
  • Description:
    Didecyldimethylammoni
  • Melting point:
    94…100 °C
  • Formula:
    C₂₂H₄₈ClN
  • Storage temperature:
    Ambient
  • Description:
    Didecyldimethylammoni
  • Boiling point:
    180 °C

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results