Analytical standard, Halfenprox, reference material, Honeywell Fluka™
Catalog # HONCR1062-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:111872-58-3
- Description:Halfenprox
- Formula:C₂₄H₂₃BrF₂O₃
- Storage temperature:2…8 °C
- Description:Halfenprox
- Boiling point:291 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results