Analytical standard, Paraquat monopyridone iodide-(1-methyl-d₃), reference material, Honeywell Fluka™
HONCR1051-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- Description:Paraquat monopyridone iodide-(1-methyl-d₃)
- Melting point:min. 300 °C
- Formula:C₁₂D₃H₁₀IN₂O
- Storage temperature:Ambient
- Description:Paraquat monopyridone iodide-(1-methyl-d₃)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results