Analytical standard, Diflovidazin, reference material, Honeywell Fluka™
Catalog # HONCR1024-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:162320-67-4
- Description:Diflovidazin
- Melting point:185 °C
- Formula:C₁₄H₇ClF₂N₄
- Storage temperature:Ambient
- Description:Diflovidazin
- Boiling point:211 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results