Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boxes
Storage Boxes
Accessories for Storage Boxes
Accessories for Storage Boxes
Accessories for Storage Boxes
Catalog # BURK3418-0001
Supplier:
BURKLE
Expand image
Accessories for Storage Boxes
Catalog # BURK3418-0001
Supplier:
BURKLE
Specifications
Description:
Lid for universal storage containers, PS, white
Seq:
04
W×D×H:
- mm
Pk:
1
Specifications
About this item
Product Details
Product Details