Order Entry
Netherlands
ContactUsLinkComponent
 
Flammable
Health
Irritant
Environment
Isohexane (mixture of isomers), PESTINORM® for pesticide residue analysis
Catalog # 85934.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Isohexane (mixture of isomers), PESTINORM® for pesticide residue analysis
Catalog # 85934.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Assay (mix. of C₆H₁₄ isomers):
    min. 99.0 %
  • 2-Methylpentane:
    min. 50.0 %
  • Identity (IR):
    Comply
  • Residue after evaporation:
    max. 0.0003 % w/w
  • Water (KF):
    max. 0.01 % w/w
  • GC/ECD any Pesticide (as Lindane):
    max. 5 pg/ml
  • Color (APHA, Pt-Co):
    max. 10

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:60 °C (1013 hPa)
Melting Pt:−154 °C
Flash Pt:−33 °C
Storage Temperature:Ambient
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :73513-42-5
Formula:C₆H₁₄
MDL Number:MFCD00009406
Melting Pt:–154 °C