Order Entry
Netherlands
ContactUsLinkComponent
 
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate 100 ±4% (by UV/Vis)
 
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate 100 ±4% (by UV/Vis)

 

  • Form:
    White powder
  • Assay (UV/vis):
    100 ± 4%
  • Reference Constants epsilonmax (pH 7, NRC):
    15.4 x 10³
  • Reference Constants epsilon260 (pH 7, DBR):
    15.4 x 10³
  • Reference Constants lambdamax (pH 7, NRC):
    259 nm
  • Storage:
    -20°C in air-tight containers
  • Spectral Ratio A250/A2:
    0.78 ± 0.03
  • Spectral Ratio A280/A260:
    0.16 ± 0.02

 

Frequently Bought Together