γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥98.5% (dry basis), Ultrapure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1132-61-2
:
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
:209.27 g/mol
Merck Index:14,06265
Melting Pt:277…280 °C (dec.)
Flash Pt:110 °C (230 °F)
EINECS:214-478-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (Dry Basis):≥98.5%
- Assay Absorbance (250 nm, 1 M, 1 cm):≤0.1
- Chloride Content:≤0.01%
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.1%
- Impurity Content Sulphate (SO₄):≤0.02%
- Loss on Drying:≤0.5%
- pKa (20 °C):7.20 ±0.15
- Iron (Fe):≤5 ppm
- Lead (Pb):≤10 ppm
Specification Test Results
Frequently Bought Together