Order Entry
Netherlands
ContactUsLinkComponent
OptoCap for optical DO sensor
OptoCap for optical DO sensor
Catalog # METN30018857
Supplier:  Mettler - Toledo
OptoCap for optical DO sensor
Catalog # METN30018857
Supplier:  Mettler - Toledo
Specifications
  • Description:
    OptoCap for optical DO sensor (InPro 6860i)
  • Pk:
    1
Specifications
About this item
The oxygen-sensitive OptoCap for the InPro 6860i sensor requires only a few seconds to replace.
  • PTFE layer avoids cell accumulation
  • Electropolished surface
  • EPDM O-ring
  • Autoclavable
Durable materials, robust design and easy replacement. Exchange of the OptoCap is recommended after a maximum 360 days. Exchange ensures high accuracy and performance over the sensor's lifetime.
Certifications: Certified to FDA/USP class VI, 3.1, N5/Ra 16.