Order Entry
Netherlands
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.9% (by GC) USP, BP, Ph. Eur., pharma grade
Dimethyl sulphoxide ≥99.9% (by GC) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide ≥99.9% (by GC) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Minimum Assay (G.C.):
    99.9%
  • Identity According to Pharmacopoeias:
    Passes Test
  • Acidity:
    Passes Test
  • Non-Volatile Matter:
    0.01%
  • Absorbance (UV):
    Passes Test
  • Related Substances (G.C.):
    0.1%
  • Substances Darkened by KOH:
    Passes Test
  • Residual Solvents (Ph.Eur/USP):
    Passes Test
  • Dimethyl Sulphone (G.C.):
    0.03%
  • Water (H₂O):
    0.1%
  • Cd, Class 1 (ICH Q3D):
    Max. 0.5 ppm
  • Pb, Class 1 (ICH Q3D):
    Max. 0.5 ppm
  • As, Class 1 (ICH Q3D):
    Max. 1.5 ppm
  • Hg, Class 1 (ICH Q3D):
    Max. 1.5 ppm
  • Co, Class 2A (ICH Q3D):
    Max. 5 ppm
  • V, Class 2A (ICH Q3D):
    Max. 10 ppm
  • Ni, Class 2A (ICH Q3D):
    Max. 20 ppm
  • Tl, Class 2B (ICH Q3D):
    Max. 0.8 ppm
  • Au, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Pd, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Ir, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Os, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Rh, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Ru, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Se, Class 2B (ICH Q3D):
    Max. 15 ppm
  • Ag, Class 2B (ICH Q3D):
    Max. 15 ppm
  • Pt, Class 2B (ICH Q3D):
    Max. 10 ppm
  • Li, Class 3 (ICH Q3D):
    Max. 55 ppm
  • Sb, Class 3 (ICH Q3D):
    Max. 120 ppm
  • Ba, Class 3 (ICH Q3D):
    Max. 140 ppm
  • Mo, Class 3 (ICH Q3D):
    Max. 25 ppm
  • Cu, Class 3 (ICH Q3D):
    Max. 250 ppm
  • Sn, Class 3 (ICH Q3D):
    Max. 600 ppm
  • Cr, Class 3 (ICH Q3D):
    Max. 25 ppm

Specification Test Results

Related Information

Frequently Bought Together