Repelcote VS water repellent
Catalog # 632478B
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Functionality:Passes test
Specification Test Results
About this item
:
Flash Pt:57 °C (closed cup)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS08