Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Catalog # VWRS24041.413
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:12,3 L
- Seq:5238
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.7 %
- Appearance of soln. (50 % V/V; water):Passes test
- IR Spectrum:Passes test
- Acidity:Max. 0.0001 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Colouration:Max. 10 APHA
- Density (20/4):1.111 - 1.115
- Substances coloured by H2SO4:Max. 150 APHA
- Formaldehyde:Max. 20 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Ignition residue (SO4):Max. 30 ppm
- Substances reducing KMnO4 (as O):Max. 3 ppm
- Water:Max. 0.1 %
- Cl (Chloride):Max. 0.2 ppm
- SO4 (Sulphate):Max. 20 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 0.5 ppm
- Pb (Lead):Max. 1 ppm
Specification Test Results
About this item
:
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-473-3
REACH:01-2119456816-28
:GHS07
CAS Number:107-21-1
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885